Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163301.2 | NP_001156773.1 | 5 | 1404 | ||
NM_001163302.2 | NP_001156774.1 | 5 | 1400 | |||
NM_001163305.2 | NP_001156777.1 | 5 | 1483 | |||
NM_001163306.2 | NP_001156778.1 | 5 | 1402 | |||
NM_001267714.1 | NP_001254643.1 | 4 | 1336 | |||
NM_001267715.1 | NP_001254644.1 | 4 | 1340 | |||
NM_008994.4 | NP_033020.2 | 6 | 1547 | |||
GenBank mRNA | AF031128.1 | - | 5 | 1395 | ||
AK146530.1 | - | 5 | 1312 | |||
AK151121.1 | - | 4 | 1234 | |||
AK152331.1 | - | 4 | 1234 | |||
BC012404.1 | - | 6 | 1427 | 3964491 |