Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166406.1 | NP_001159878.1 | 2 | 372 | ||
NM_001166407.1 | NP_001159879.1 | 2 | 370 | |||
NM_009004.4 | NP_033030.1 | 2 | 266 | |||
GenBank mRNA | AK051201.1 | - | 2 | 184 | ||
AK051995.1 | - | 2 | 221 | |||
AK053280.1 | - | 2 | 184 | |||
AK083412.1 | - | 2 | 226 | |||
AK083697.1 | - | 2 | 186 | |||
AK084732.1 | - | 2 | 208 | |||
AK142764.1 | - | 2 | 196 | |||
AK144844.1 | - | 2 | 184 | |||
BC060608.1 | - | 2 | 372 | 5720408 | ||
BY012551.1 | - | 2 | 270 | |||
Y09632.1 | - | 2 | 421 |