Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293652.1 | NP_001280581.1 | 6 | 674 | ||
NM_001293653.1 | NP_001280582.1 | 4 | 335 | |||
NM_001293654.1 | NP_001280583.1 | 5 | 501 | |||
NM_001293655.1 | NP_001280584.1 | 4 | 414 | |||
NM_009005.3 | NP_033031.2 | 4 | 452 | |||
GenBank mRNA | AB232599.1 | - | 3 | 246 | ||
AK005004.1 | - | 4 | 414 | |||
AK057671.1 | - | 4 | 381 | |||
AK135659.1 | - | 5 | 502 | |||
AK139914.1 | - | 4 | 405 | |||
AK148117.1 | - | 4 | 445 | |||
AK151078.1 | - | 4 | 426 | |||
AK152666.1 | - | 6 | 649 | |||
AK154234.1 | - | 4 | 433 | |||
AK158971.1 | - | 4 | 336 | |||
AK159507.1 | - | 4 | 381 | |||
AK159928.1 | - | 4 | 353 | |||
BC004597.1 | - | 4 | 370 | 3584891 | ||
BC086793.1 | - | 4 | 368 | 6414012 | ||
CT010352.1 | - | 3 | 246 | |||
X89650.1 | - | 4 | 402 |