Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009009.4 | NP_033035.3 | 14 | 2511 | ||
GenBank mRNA | AK004746.1 | - | 14 | 2314 | ||
AK128984.1 | - | 14 | 2400 | |||
AK129050.1 | - | 9 | 1639 | |||
AK139492.1 | - | 13 | 2164 | |||
AK145591.1 | - | 14 | 2478 | |||
AK163992.1 | - | 2372 | ||||
AK168718.1 | - | 14 | 2499 | |||
AK168896.1 | - | 14 | 2460 | |||
BC043032.1 | - | 13 | 2280 | 5696540 | ||
BC129916.1 | - | 13 | 2310 | 40111820 | ||
BC129917.1 | - | 13 | 2311 | 40111821 |