Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009037.2 | NP_033063.1 | 1 | 302 | ||
GenBank mRNA | AK017494.1 | - | 1 | 209 | ||
AK133971.1 | - | 1 | 252 | |||
AK159973.1 | - | 1 | 205 | |||
BC049108.1 | - | 1 | 195 | 6414128 | ||
D13003.1 | - | 1 | 222 |