Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177767.1 | NP_001171238.1 | 5 | 539 | ||
GenBank mRNA | AF051348.1 | - | 534 | |||
BC067397.1 | - | 3 | 294 | 30462971 | ||
CD542867.1 | - | 5 | 523 | |||
CF162675.1 | - | 5 | 523 |
Gene Symbol: Gm13242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103158.2 | NP_001096628.2 | 5 | 539 | ||
GenBank mRNA | BC096593.1 | - | 5 | 551 | 30459933 |
Gene Symbol: Zfp600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177545.2 | NP_001171016.1 | 5 | 446 | ||
NM_001177546.2 | NP_001171017.1 | 5 | 539 | |||
GenBank mRNA | BC108352.1 | - | 5 | 446 | 30462028 | |
CD539901.1 | - | 5 | 539 |