Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080127.1 | NP_001073596.1 | 3 | 274 | ||
NM_001080128.1 | NP_001073597.1 | 2 | 93 | |||
NM_009070.2 | NP_033096.2 | 4 | 396 | |||
GenBank mRNA | AK011475.1 | - | 3 | 276 | ||
AK146443.1 | - | 2 | 92 | |||
AK151150.1 | - | 3 | 264 | |||
AK165905.1 | - | 2 | 80 | |||
AK178555.1 | - | 2 | 225 | |||
AK182154.1 | - | 1 | 44 | |||
AK206201.1 | - | 2 | 230 | |||
AK211825.1 | - | 1 | 44 | |||
AK213636.1 | - | 1 | 44 | |||
BC002061.1 | - | 3 | 262 | 3485311 | ||
BY013195.1 | - | 2 | 94 | |||
CJ058744.1 | - | 3 | 276 | |||
X70067.1 | - | 3 | 284 |