Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243211.1 | NP_001230140.1 | 3 | 397 | ||
NM_001562.3 | NP_001553.1 | 4 | 409 | |||
XM_011542805.1 | XP_011541107.1 | 3 | 405 | |||
XM_011542806.2 | XP_011541108.1 | 4 | 413 | |||
GenBank mRNA | AF380360.1 | - | 4 | 241 | ||
AY044641.1 | - | 4 | 286 | |||
AY266351.1 | - | 3 | 354 | |||
BC007007.1 | - | 4 | 384 | 3686124 | ||
BC007461.1 | - | 4 | 409 | 3681009 | ||
CR541973.1 | - | 3 | 189 | |||
CR542001.1 | - | 3 | 189 | |||
D49950.1 | - | 4 | 366 | |||
U90434.1 | - | 3 | 186 |