Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009099.2 | NP_033125.2 | 2 | 305 | ||
GenBank mRNA | AF220014.1 | - | 2 | 222 | ||
AF220015.1 | - | 2 | 222 | |||
AK089726.1 | - | 2 | 302 | |||
AK134144.1 | - | 2 | 274 | |||
AK137506.1 | - | 2 | 306 | |||
AK138249.1 | - | 2 | 308 | |||
BC005447.1 | - | 2 | 239 | 3493256 | ||
J03776.1 | - | 2 | 222 |