Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009030.3 | NP_033056.2 | 3 | 317 | ||
GenBank mRNA | AK082419.1 | - | 3 | 289 | ||
AK141176.1 | - | 2 | 771 | |||
AK166437.1 | - | 3 | 287 | |||
BC138568.1 | - | 3 | 295 | 8861588 | ||
BC138570.1 | - | 3 | 296 | 8861590 | ||
BY277283.1 | - | 3 | 317 | |||
U35141.1 | - | 3 | 287 |