Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080927.2 | NP_001074396.1 | 5 | 674 | ||
NM_001080928.1 | NP_001074397.1 | 5 | 560 | |||
NM_001277116.1 | NP_001264045.1 | 7 | 756 | |||
NM_009035.5 | NP_033061.3 | 5 | 576 | |||
GenBank mRNA | AK028363.1 | - | 7 | 734 | ||
AK080359.1 | - | 5 | 469 | |||
AK090006.1 | - | 5 | 462 | |||
AK132145.1 | - | 5 | 518 | |||
AK139087.1 | - | 5 | 461 | |||
AK145187.1 | - | 5 | 468 | |||
AK152746.1 | - | 5 | 576 | |||
AK153163.1 | - | 6 | 464 | |||
BC035299.1 | - | 3 | 362 | |||
BC051387.1 | - | 5 | 505 | 4460290 | ||
CD540070.1 | - | 5 | 514 | |||
S63463.1 | - | 5 | 577 | |||
X17459.1 | - | 5 | 572 | |||
Y18248.1 | - | 2 | 189 |