Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcvrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009038.2 | NP_033064.1 | 1 | 351 | ||
GenBank mRNA | BC110564.2 | - | 1 | 344 | 40050126 | |
CK617354.1 | - | 1 | 311 | |||
X66196.1 | - | 1 | 359 |