Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ret | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080780.1 | NP_001074249.1 | 4 | 1339 | ||
NM_009050.2 | NP_033076.2 | 4 | 1339 | |||
GenBank mRNA | AF209436.1 | - | 4 | 728 | ||
AK044686.1 | - | 4 | 926 | |||
AK051633.1 | - | 4 | 918 | |||
AK084977.1 | - | 4 | 988 | |||
AY326397.1 | - | 4 | 728 | |||
BC059012.1 | - | 4 | 1313 | 6849696 | ||
X67812.1 | - | 4 | 728 |