Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009066.3 | NP_033092.3 | 5 | 783 | ||
GenBank mRNA | AK154962.1 | - | 5 | 744 | ||
AK155509.1 | - | 5 | 759 | |||
BC009070.1 | - | 5 | 746 | 2651194 | ||
BC082771.1 | - | 2 | 1254 | |||
CT010219.1 | - | 3 | 473 | |||
CV560731.1 | - | 3 | 416 | |||
KJ650216.1 | - | 4 | 560 | |||
Y12881.1 | - | 3 | 479 |