Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080127.1 | NP_001073596.1 | 3 | 343 | ||
NM_001080128.1 | NP_001073597.1 | 2 | 162 | |||
NM_009070.2 | NP_033096.2 | 4 | 465 | |||
GenBank mRNA | AK011475.1 | - | 3 | 345 | ||
AK146443.1 | - | 2 | 161 | |||
AK151150.1 | - | 3 | 333 | |||
AK165905.1 | - | 2 | 149 | |||
AK182154.1 | - | 1 | 113 | |||
AK206201.1 | - | 2 | 299 | |||
AK211825.1 | - | 1 | 113 | |||
AK213636.1 | - | 1 | 113 | |||
BB868062.1 | - | 3 | 287 | |||
BC002061.1 | - | 3 | 331 | 3485311 | ||
BY013195.1 | - | 2 | 163 | |||
CJ058744.1 | - | 3 | 345 | |||
X70067.1 | - | 3 | 353 |