Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288705.1 | NP_001275634.1 | 11 | 1892 | ||
NM_005211.3 | NP_005202.2 | 12 | 1971 | |||
NR_109969.1 | 11 | 1892 | ||||
GenBank mRNA | AB208819.1 | - | 11 | 1842 | ||
AK304491.1 | - | 11 | 1673 | |||
BC047521.1 | - | 11 | 1874 | 4819527 | ||
BX648599.1 | - | 10 | 1762 | |||
X03663.1 | - | 12 | 1979 |