Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277113.1 | NP_001264042.1 | 3 | 341 | ||
NM_009079.3 | NP_033105.1 | 3 | 391 | |||
GenBank mRNA | AA755840.1 | - | 3 | 393 | ||
AK003022.1 | - | 3 | 257 | |||
AK009094.1 | - | 3 | 258 | |||
AK168353.1 | - | 3 | 255 | |||
AK169078.1 | - | 3 | 244 | |||
AK169731.1 | - | 3 | 257 | |||
BC007139.1 | - | 3 | 264 | 3487607 | ||
BC021344.1 | - | 3 | 250 | 4190569 | ||
BC096390.1 | - | 3 | 400 | 1179671 | ||
D17653.1 | - | 3 | 255 |