Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009090.5 | NP_033116.2 | 5 | 431 | ||
GenBank mRNA | AK137796.1 | - | 364 | |||
AK146763.1 | - | 5 | 361 | |||
AK151270.1 | - | 5 | 359 | |||
AK160053.1 | - | 5 | 366 | |||
BC002023.1 | - | 5 | 349 | 3492868 | ||
BC099548.1 | - | 5 | 376 | 30312083 | ||
D83999.1 | - | 5 | 352 |