Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009091.2 | NP_033117.1 | 3, 4 | 370 | ||
GenBank mRNA | AA027738.1 | - | 2, 3 | 297 | ||
AK135319.1 | - | 3, 4 | 344 | |||
AK146147.1 | - | 3, 4 | 342 | |||
AV569793.1 | - | 3, 4 | 370 | |||
BC010763.1 | - | 2, 3 | 336 | 4234356 | ||
BC094409.1 | - | 2, 3 | 335 | 1179896 | ||
M33330.1 | - | 3, 4 | 350 |