Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009094.1 | NP_033120.1 | 2 | 244 | ||
GenBank mRNA | AK002445.1 | - | 3 | 262 | ||
AK010624.1 | - | 3 | 265 | |||
AK010676.1 | - | 3 | 262 | |||
AK010921.1 | - | 3 | 262 | |||
AK012429.1 | - | 3 | 262 | |||
AK014210.1 | - | 3 | 260 | |||
AK088410.1 | - | 3 | 262 | |||
AK135369.1 | - | 3 | 265 | |||
AK153211.1 | - | 3 | 256 | |||
BC009100.1 | - | 2 | 269 | 3482299 | ||
BC089348.1 | - | 2 | 246 | |||
BC089349.1 | - | 2 | 240 | |||
BC106152.1 | - | 2 | 245 | 3669367 | ||
M73436.1 | - | 2 | 244 |