Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159725.2 | NP_001153197.2 | 6 | 1325 | ||
NM_008998.4 | NP_033024.1 | 6 | 1556 | |||
GenBank mRNA | AK009707.1 | - | 6 | 1326 | ||
AK075591.1 | - | 6 | 1556 | |||
BC013170.1 | - | 6 | 1405 | 2648145 | ||
BC051071.1 | - | 6 | 1532 | 6307319 |