Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297606.1 | NP_001284535.1 | 2 | 195 | ||
NM_001297607.1 | NP_001284536.1 | 2 | 195 | |||
NM_009010.5 | NP_033036.2 | 2 | 195 | |||
GenBank mRNA | AK038300.1 | - | 2 | 172 | ||
AK045224.1 | - | 2 | 185 | |||
AK161875.1 | - | 2 | 172 | |||
AK165467.1 | - | 2 | 196 | |||
BC003846.1 | - | 2 | 183 | |||
BC138684.1 | - | 2 | 155 | 8861705 | ||
BC145372.1 | - | 2 | 155 | 9053930 | ||
X92410.1 | - | 2 | 95 |