Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009030.3 | NP_033056.2 | 3, 4 | 452 | ||
GenBank mRNA | AK082419.1 | - | 3, 4 | 424 | ||
AK166437.1 | - | 3, 4 | 422 | |||
BC138568.1 | - | 3, 4 | 430 | 8861588 | ||
BC138570.1 | - | 3, 4 | 431 | 8861590 | ||
U35141.1 | - | 3, 4 | 422 |