Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Relb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290457.1 | NP_001277386.1 | 10 | 1433 | ||
NM_009046.2 | NP_033072.2 | 11 | 1442 | |||
GenBank mRNA | AK146932.1 | - | 11 | 1443 | ||
AK156767.1 | - | 11 | 1398 | |||
BC019765.1 | - | 11 | 1442 | 3584200 | ||
BC034523.1 | - | 10 | 1246 | 4485540 | ||
BC117793.1 | - | 11 | 1378 | 40092162 | ||
DQ020178.1 | - | 10 | 1094 | |||
DQ020179.1 | - | 10 | 2210 | |||
DQ020180.1 | - | 12 | 2250 | |||
DQ020181.1 | - | 11 | 2316 | |||
M83380.1 | - | 11 | 1279 |