Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009047.5 | NP_033073.1 | 5 | 1293 | ||
GenBank mRNA | AK033315.1 | - | 64 | |||
AK053133.1 | - | 5 | 1294 | |||
AK078772.1 | - | 64 | ||||
AK084460.1 | - | 5 | 1301 |