Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009055.4 | NP_033081.3 | 21 | 3299 | ||
GenBank mRNA | AK037226.1 | - | 14 | 2242 | ||
AK083571.1 | - | 21 | 3270 | |||
AK147790.1 | - | 21 | 3340 | |||
AK196928.1 | - | 1 | 90 | |||
BC057018.1 | - | 21 | 3278 | 5705637 |