Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009059.2 | NP_033085.2 | 9 | 1373 | ||
GenBank mRNA | AK144440.1 | - | 9 | 1375 | ||
AK204851.1 | - | 2 | 114 | |||
AK215408.1 | - | 2 | 159 | |||
BC068121.1 | - | 9 | 1373 | 6415928 | ||
KJ650210.1 | - | 8 | 1181 | |||
U54639.1 | - | 9 | 1302 |