Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004474.3 | NP_004465.3 | 1 | 3353 | ||
GenBank mRNA | AF042832.1 | - | 1 | 3367 |