Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009107.3 | NP_033133.1 | 2 | 663 | ||
GenBank mRNA | BC058401.1 | - | 2 | 645 | 5707723 | |
BM947610.1 | - | 2 | 389 | |||
G49237.1 | - | 2 | 395 | |||
M84819.1 | - | 2 | 308 | |||
S62948.1 | - | 2 | 507 | |||
X66225.1 | - | 2 | 395 |