Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009127.4 | NP_033153.2 | 6 | 1678 | ||
GenBank mRNA | AF509567.1 | - | 6 | 1513 | ||
AF509570.1 | - | 6 | 1513 | |||
AK135642.1 | - | 3 | 867 | |||
BC007474.1 | - | 6 | 1522 | 3599932 | ||
BQ885468.1 | - | 1 | 514 |