Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042613.1 | NP_001036078.1 | 2 | 147 | ||
NM_001042614.1 | NP_001036079.1 | 2 | 230 | |||
NM_009155.3 | NP_033181.3 | 2 | 164 | |||
GenBank mRNA | AI892015.1 | - | 2 | 239 | ||
AK002450.1 | - | 2 | 146 | |||
AK077265.1 | - | 2 | 146 | |||
AK146774.1 | - | 2 | 146 | |||
AK148623.1 | - | 2 | 146 | |||
AK159269.1 | - | 2 | 146 | |||
AK159524.1 | - | 2 | 147 | |||
AK159894.1 | - | 2 | 143 | |||
AK159911.1 | - | 2 | 147 | |||
AK159935.1 | - | 2 | 146 | |||
AK167611.1 | - | 2 | 146 | |||
AK168463.1 | - | 2 | 143 | |||
AK168583.1 | - | 2 | 143 | |||
AK168631.1 | - | 2 | 146 | |||
AK168747.1 | - | 2 | 146 | |||
AK168926.1 | - | 2 | 146 | |||
AK169011.1 | - | 2 | 145 | |||
AK169158.1 | - | 2 | 145 | |||
AK169362.1 | - | 2 | 146 | |||
AK188386.1 | - | 2 | 131 | |||
BC001991.1 | - | 2 | 147 | 3486761 | ||
CB231357.1 | - | 2 | 167 | |||
CK627189.1 | - | 2 | 147 | |||
X99807.1 | - | 1 | 207 |