Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009160.2 | NP_033186.1 | 2, 3 | 271 | ||
GenBank mRNA | BC003705.1 | - | 2, 3 | 222 | 3601620 | |
BC012896.1 | - | 2, 3 | 219 | |||
BY116790.1 | - | 2, 3 | 273 | |||
L40156.1 | - | 1, 2 | 199 |