Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009164.2 | NP_033190.2 | 7 | 686 | ||
GenBank mRNA | AK145156.1 | - | 7 | 626 | ||
AK161215.1 | - | 6 | 610 | |||
AK166337.1 | - | 7 | 679 | |||
AK172060.1 | - | 7 | 683 | |||
BC004598.1 | - | 5 | 407 | 3584958 | ||
X87671.1 | - | 4 | 542 |