Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034962.1 | NP_001030134.1 | 5 | 338 | ||
NM_001034963.1 | NP_001030135.1 | 4 | 248 | |||
NM_001034964.1 | NP_001030136.1 | 3 | 163 | |||
NM_009166.3 | NP_033192.2 | 6 | 527 | |||
NM_178362.1 | NP_848139.1 | 5 | 614 | |||
GenBank mRNA | AF078666.1 | - | 4 | 237 | ||
AF078667.1 | - | 5 | 327 | |||
AF521593.1 | - | 2 | 120 | |||
AK035212.1 | - | 5 | 330 | |||
AK087553.1 | - | 5 | 614 | |||
AK122487.2 | - | 5 | 282 | |||
AK146712.1 | - | 3 | 166 | |||
AK166943.1 | - | 4 | 305 | |||
AK216997.1 | - | 1 | 18 | |||
BC012703.1 | - | 5 | 327 | 3602997 | ||
BY031726.1 | - | 5 | 302 | |||
U58883.1 | - | 6 | 465 |