Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159745.1 | NP_001153217.1 | 2 | 549 | ||
NM_009183.2 | NP_033209.1 | 2 | 549 | |||
GenBank mRNA | AJ223956.1 | - | 2 | 540 | ||
AK003690.1 | - | 2 | 491 | |||
AK041723.1 | - | 2 | 482 | |||
AK165717.1 | - | 2 | 524 | |||
BC060112.1 | - | 2 | 526 | 6405078 | ||
X86000.1 | - | 549 |