Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159529.1 | NP_001153001.1 | 3, 4 | 339 | ||
NM_009187.3 | NP_033213.1 | 2, 3 | 270 | |||
GenBank mRNA | AK009614.1 | - | 3, 4 | 336 | ||
AK133520.1 | - | 2, 3 | 270 | |||
AK150034.1 | - | 2, 3 | 267 | |||
AK152553.1 | - | 2, 3 | 260 | |||
AK159441.1 | - | 2, 3 | 268 | |||
AK166761.1 | - | 2, 3 | 255 | |||
AK167062.1 | - | 2, 3 | 265 | |||
AK184659.1 | - | 1, 2 | 145 | |||
AK216357.1 | - | 1, 2 | 145 | |||
BY122624.1 | - | 2, 3 | 270 | |||
X80899.1 | - | 2, 3 | 238 |