Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009190.2 | NP_033216.2 | 2 | 370 | ||
GenBank mRNA | AK151023.1 | - | 2 | 324 | ||
AK152131.1 | - | 2 | 357 | |||
AK165594.1 | - | 315 | ||||
AK168270.1 | - | 2 | 261 | |||
BC003799.1 | - | 2 | 277 | 3488471 | ||
U10119.1 | - | 2 | 321 |