Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009200.3 | NP_033226.1 | 5, 6 | 768 | ||
GenBank mRNA | AK159197.1 | - | 5, 6 | 769 | ||
BC138120.1 | - | 5, 6 | 670 | 8861139 | ||
BC138140.1 | - | 5, 6 | 670 | 8861159 | ||
D83262.1 | - | 5, 6 | 762 |