Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009202.5 | NP_033228.2 | 3 | 818 | ||
GenBank mRNA | AF010259.1 | - | 3 | 676 | ||
BC021651.1 | - | 3 | 818 | 4211979 | ||
U38652.1 | - | 3 | 673 |