Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009101.2 | NP_033127.1 | 4 | 446 | ||
GenBank mRNA | AK155855.1 | - | 4 | 476 | ||
BC009105.1 | - | 4 | 463 | 3483313 | ||
BC110562.1 | - | 4 | 448 | 40049793 | ||
BC110563.1 | - | 4 | 448 | 40049794 | ||
BC145932.1 | - | 4 | 492 | 40131235 | ||
BC145934.1 | - | 4 | 492 | 40131237 | ||
M21019.1 | - | 4 | 448 |