Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009140.2 | NP_033166.1 | 3 | 270 | ||
GenBank mRNA | AK137628.1 | - | 3 | 298 | ||
AK150450.1 | - | 3 | 178 | |||
AK155458.1 | - | 1 | 506 | |||
AK155874.1 | - | 3 | 299 | |||
AK155916.1 | - | 3 | 297 | |||
AK157079.1 | - | 3 | 297 | |||
BC119511.1 | - | 3 | 244 | 40055368 | ||
BC119512.1 | - | 3 | 244 | 40055369 | ||
X53798.1 | - | 3 | 270 |