Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009143.3 | NP_033169.3 | 3 | 655 | ||
GenBank mRNA | AK002496.1 | - | 3 | 411 | ||
BC058798.1 | - | 3 | 655 | 6485217 | ||
BC062881.1 | - | 3 | 401 | 5693309 | ||
BC087871.1 | - | 3 | 300 | 30279332 | ||
D50646.1 | - | 3 | 409 |