Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113478.1 | NP_001106950.1 | 4 | 409 | ||
NM_009146.3 | NP_033172.2 | 4 | 371 | |||
GenBank mRNA | AK075573.1 | - | 4 | 363 | ||
AK144933.1 | - | 4 | 363 | |||
AK162693.1 | - | 4 | 364 | |||
AK167289.1 | - | 4 | 399 | |||
AK213195.1 | - | 2 | 62 | |||
AU067508.1 | - | 4 | 371 | |||
BC027770.1 | - | 3 | 369 | 5344922 | ||
D50464.1 | - | 4 | 359 |