Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009154.2 | NP_033180.2 | 4 | 862 | ||
GenBank mRNA | AK031231.1 | - | 4 | 855 | ||
AK039101.1 | - | 4 | 848 | |||
AK043386.1 | - | 4 | 574 | |||
AK045236.1 | - | 4 | 866 | |||
AK046502.1 | - | 4 | 638 | |||
AK141612.1 | - | 4 | 806 | |||
AK143026.1 | - | 4 | 873 | |||
AK157706.1 | - | 4 | 845 | |||
AK165206.1 | - | 4 | 1163 | |||
BC065137.1 | - | 3 | 331 | 30548165 | ||
X97817.1 | - | 2 | 231 |