Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042613.1 | NP_001036078.1 | 2 | 218 | ||
NM_001042614.1 | NP_001036079.1 | 2 | 301 | |||
NM_009155.3 | NP_033181.3 | 2 | 235 | |||
GenBank mRNA | AI892015.1 | - | 2 | 310 | ||
AK002450.1 | - | 2 | 217 | |||
AK077265.1 | - | 2 | 217 | |||
AK146774.1 | - | 2 | 217 | |||
AK148623.1 | - | 2 | 217 | |||
AK159269.1 | - | 2 | 217 | |||
AK159524.1 | - | 2 | 218 | |||
AK159894.1 | - | 2 | 214 | |||
AK159911.1 | - | 2 | 218 | |||
AK159935.1 | - | 2 | 217 | |||
AK167611.1 | - | 2 | 217 | |||
AK168463.1 | - | 2 | 214 | |||
AK168583.1 | - | 2 | 214 | |||
AK168631.1 | - | 2 | 217 | |||
AK168747.1 | - | 2 | 217 | |||
AK168926.1 | - | 2 | 217 | |||
AK169011.1 | - | 2 | 216 | |||
AK169158.1 | - | 2 | 216 | |||
AK169362.1 | - | 2 | 217 | |||
AK190911.1 | - | 1 | 52 | |||
AK192055.1 | - | 1 | 52 | |||
AK192611.1 | - | 1 | 52 | |||
AK193239.1 | - | 1 | 52 | |||
AK193304.1 | - | 1 | 52 | |||
AK193589.1 | - | 1 | 52 | |||
AK196001.1 | - | 1 | 52 | |||
AK199796.1 | - | 1 | 52 | |||
AK200678.1 | - | 1 | 52 | |||
AK201439.1 | - | 1 | 52 | |||
AK201752.1 | - | 1 | 52 | |||
AK203751.1 | - | 1 | 52 | |||
BC001991.1 | - | 2 | 218 | 3486761 | ||
CB231357.1 | - | 2 | 238 | |||
CK627189.1 | - | 2 | 218 | |||
X99807.1 | - | 1 | 278 |