Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034962.1 | NP_001030134.1 | 5 | 406 | ||
NM_001034963.1 | NP_001030135.1 | 4 | 316 | |||
NM_001034964.1 | NP_001030136.1 | 3 | 231 | |||
NM_009166.3 | NP_033192.2 | 6 | 595 | |||
NM_178362.1 | NP_848139.1 | 5 | 682 | |||
GenBank mRNA | AF078666.1 | - | 4 | 305 | ||
AF078667.1 | - | 5 | 395 | |||
AF521593.1 | - | 2 | 188 | |||
AK035212.1 | - | 5 | 398 | |||
AK087553.1 | - | 5 | 682 | |||
AK122487.2 | - | 5 | 350 | |||
AK146712.1 | - | 3 | 234 | |||
AK166943.1 | - | 4 | 373 | |||
AK208107.1 | - | 1 | 60 | |||
BC012703.1 | - | 5 | 395 | 3602997 | ||
U58883.1 | - | 6 | 533 |