Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005730.3 | NP_005721.3 | 2 | 654 | ||
XM_005268556.1 | XP_005268613.1 | 2 | 414 | |||
GenBank mRNA | AF000152.1 | - | 2 | 430 | ||
AF022231.1 | - | 2 | 371 | |||
AK226076.1 | - | 2 | 410 | |||
AK291289.1 | - | 2 | 401 | |||
AK310873.1 | - | 2 | 156 | |||
AK312413.1 | - | 2 | 413 | |||
AY279531.1 | - | 2 | 125 | |||
BC018922.2 | - | 2 | 386 | |||
BC065920.1 | - | 2 | 369 | 5756196 | ||
CN313995.1 | - | 2 | 579 |