Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289725.1 | NP_001276654.1 | 2, 3 | 505 | ||
NM_009193.2 | NP_033219.1 | 3, 4 | 610 | |||
GenBank mRNA | AK131729.1 | - | 381 | |||
AK154053.1 | - | 3, 4 | 357 | |||
AK162121.1 | - | 3, 4 | 356 | |||
AK210098.1 | - | 2, 3 | 149 | |||
BC025113.1 | - | 3, 4 | 345 | |||
U75680.1 | - | 3, 4 | 342 |