Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009194.3 | NP_033220.2 | 2 | 960 | ||
GenBank mRNA | U13174.1 | - | 2 | 957 |