Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253804.1 | NP_001240733.1 | 2 | 262 | ||
NM_009195.3 | NP_033221.1 | 2 | 256 | |||
GenBank mRNA | AF047339.1 | - | 2 | 180 | ||
AF121118.1 | - | 2 | 247 | |||
AK149969.1 | - | 2 | 245 | |||
AK151717.1 | - | 2 | 217 | |||
AK152762.1 | - | 2 | 234 | |||
AK153245.1 | - | 2 | 234 | |||
AK159274.1 | - | 2 | 244 | |||
AK159331.1 | - | 2 | 239 | |||
AK159483.1 | - | 2 | 212 | |||
AK159537.1 | - | 2 | 239 | |||
AK160034.1 | - | 2 | 205 | |||
AK160072.1 | - | 2 | 239 | |||
AK164479.1 | - | 2 | 221 | |||
AK165874.1 | - | 2 | 234 | |||
AK166304.1 | - | 2 | 215 | |||
AK167371.1 | - | 2 | 253 | |||
AK167444.1 | - | 2 | 253 | |||
AK169044.1 | - | 2 | 211 | |||
BC066872.1 | - | 2 | 186 | 30436525 | ||
BY740705.1 | - | 2 | 257 |